Gene Transcripts

Gene Symbol
Hist1h2bh
Entrez Gene ID
319182
Gene Name
histone cluster 1, H2bh
Gene Aliases
H2b-221
Chromosome Location
Chr. 13: 23542923 - 23543444 on Build GRCm38
UniGene ID
Mm.352273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178197.2 NP_835504.1 1 168
GenBank mRNA AK158416.1 - 117
BC092138.1 - 83 30613720
BC132584.1 - 1 168 40130861
BC132586.1 - 1 168 40130863

Gene Symbol
Hist1h2bc
Entrez Gene ID
68024
Gene Name
histone cluster 1, H2bc
Gene Aliases
2610022J01Rik, H2bfs, R74621
Chromosome Location
Chr. 13: 23684199 - 23692480 on Build GRCm38
UniGene ID
Mm.261676
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290380.1 NP_001277309.1 1 109
NM_023422.3 NP_075911.2 1 109
GenBank mRNA AK005191.1 - 1 104
AK005407.1 - 1 104
AK011516.1 - 1 111
BC019673.1 - 1 92 3993954
BC082774.1 - 1 92

Gene Symbol
Hist1h2be
Entrez Gene ID
319179
Gene Name
histone cluster 1, H2be
Gene Aliases
Chromosome Location
Chr. 13: 23583670 - 23621124 on Build GRCm38
UniGene ID
Mm.440391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177653.1 NP_001171124.1 2 269
NM_001290530.1 NP_001277459.1 1 90
NM_178194.4 NP_835501.1 2 295
GenBank mRNA AK030546.1 - 2 297
AK049948.1 - 2 270
BC069889.1 - 1 83 4217814

Gene Symbol
Hist1h2bf
Entrez Gene ID
319180
Gene Name
histone cluster 1, H2bf
Gene Aliases
Chromosome Location
Chr. 13: 23573760 - 23574190 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178195.2 NP_835502.1 1 76

Gene Symbol
Hist1h2bg
Entrez Gene ID
319181
Gene Name
histone cluster 1, H2bg
Gene Aliases
Chromosome Location
Chr. 13: 23571400 - 23571863 on Build GRCm38
UniGene ID
Mm.261673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178196.4 NP_835503.1 1 109
GenBank mRNA BC060304.1 - 1 136

Gene Symbol
Hist1h2bj
Entrez Gene ID
319183
Gene Name
histone cluster 1, H2bj
Gene Aliases
Chromosome Location
Chr. 13: 22043230 - 22043658 on Build GRCm38
UniGene ID
Mm.371766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178198.2 NP_835505.1 1 76
GenBank mRNA BC115792.1 - 68

Gene Symbol
Hist1h2bk
Entrez Gene ID
319184
Gene Name
histone cluster 1, H2bk
Gene Aliases
Chromosome Location
Chr. 13: 22035821 - 22036320 on Build GRCm38
UniGene ID
Mm.358955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175665.2 NP_783596.1 1 142
GenBank mRNA BC147143.1 - 1 142
BC147144.1 - 1 142 9056197

Gene Symbol
Hist1h2bl
Entrez Gene ID
319185
Gene Name
histone cluster 1, H2bl
Gene Aliases
H2b-291a
Chromosome Location
Chr. 13: 21715713 - 21716143 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178199.2 NP_835506.1 1 76

Gene Symbol
Hist1h2bn
Entrez Gene ID
319187
Gene Name
histone cluster 1, H2bn
Gene Aliases
Chromosome Location
Chr. 13: 21754123 - 21754553 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178201.2 NP_835508.1 1 76

Gene Symbol
Hist1h2bq
Entrez Gene ID
665596
Gene Name
histone cluster 1, H2bq
Gene Aliases
Gm13646, OTTMUSG00000013203
Chromosome Location
Chr. 13: 21806412 - 21810199 on Build GRCm38
UniGene ID
Mm.371766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097979.2 NP_001091448.2 1 104

Gene Symbol
Hist1h2br
Entrez Gene ID
665622
Gene Name
histone cluster 1 H2br
Gene Aliases
Gm11277, OTTMUSG00000000449
Chromosome Location
Chr. 13: 21833743 - 21837530 on Build GRCm38
UniGene ID
Mm.471247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110555.1 NP_001104025.1 1 104
GenBank mRNA AK036389.1 - 1 105
BC011440.1 - 1 89 3989862

View Full Product Details