Gene Transcripts

Gene Symbol
IQCE
Entrez Gene ID
23288
Gene Name
IQ motif containing E
Gene Aliases
1700028P05Rik
Chromosome Location
Chr. 7: 2558972 - 2614734 on Build GRCh38
UniGene ID
Hs.520627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287501.1 NP_001274430.1 20 2152
NM_152558.4 NP_689771.3 22 2246
XM_017011902.1 XP_016867391.1 22 2308
XM_017011903.1 XP_016867392.1 22 2186
GenBank mRNA AB028946.2 - 20 1949
AL136792.1 - 21 1927
BC043150.1 - 20 2072 5297871
BC071858.1 - 22 2220 5478116
DA246430.1 - 5 424
KJ534872.1 - 22 2036
KJ534873.1 - 6 320
KJ535081.1 - 20 1942

View Full Product Details