Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003750319.3 | XP_003750367.1 | 23 | 2933 | |
XM_006241324.2 | XP_006241386.1 | 22 | 3929 | ||
XM_006241325.1 | XP_006241387.1 | 21 | 2500 | ||
XM_006241326.2 | XP_006241388.1 | 21 | 3892 | ||
XM_008765365.1 | XP_008763587.1 | 23 | 2652 | ||
XM_008765367.1 | XP_008763589.1 | 22 | 9357 | ||
GenBank mRNA | BC168899.1 | - | 4 | 804 |