Gene Transcripts

Gene Symbol
Snrpg
Entrez Gene ID
681031
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
Chromosome Location
Chr. 4: 118167294 - 118174843 on Build Rnor_6.0
UniGene ID
Rn.2397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135085.1 NP_001128557.1 4 343
GenBank mRNA BC168750.1 - 4 309
FQ217038.1 - 4 309
FQ221105.1 - 4 341
FQ221517.1 - 4 309
FQ221580.1 - 4 370
FQ221827.1 - 4 311
FQ222397.1 - 4 309
FQ222427.1 - 4 312
FQ222653.1 - 4 358
FQ223239.1 - 4 309
FQ223409.1 - 4 358
FQ228483.1 - 4 312
FQ228634.1 - 4 317
FQ228635.1 - 4 354

Gene Symbol
LOC687679
Entrez Gene ID
687679
Gene Name
similar to small nuclear ribonucleoprotein polypeptide G
Gene Aliases
Chromosome Location
Chr. 1: 91588215 - 91588610 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748839.2 XP_003748887.1 1 310

View Full Product Details