Gene Transcripts

Gene Symbol
Sult2a6
Entrez Gene ID
24902
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
Gene Aliases
RATSMP2A, RATSMP2B, Smp2a, St2a2, Sult2a4l1, Sult2al1
Chromosome Location
Chr. 1: 76529121 - 76722965 on Build Rnor_6.0
UniGene ID
Rn.221887
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012695.3 NP_036827.3 4, 5 609
GenBank mRNA FQ209484.1 - 4, 5 484
FQ209563.1 - 4, 5 484
FQ210641.1 - 3, 4 357
FQ219267.1 - 4, 5 484
FQ219345.1 - 4, 5 484
M33329.1 - 4, 5 609
X63410.1 - 4, 5 609

Gene Symbol
Sult2a1
Entrez Gene ID
24912
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
Gene Aliases
STa, Smp-2, St2, St2a1, Sth2
Chromosome Location
Chr. 1: 76558721 - 76614309 on Build Rnor_6.0
UniGene ID
Rn.91378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131903.1 NP_571978.1 4, 5 609
XM_008758891.1 XP_008757113.1 4, 5 639
XM_008758892.1 XP_008757114.1 4, 5 638
XM_008758893.1 XP_008757115.1 4, 5 627
GenBank mRNA D14987.1 - 4, 5 609
D14988.1 - 4, 5 609
FQ209430.1 - 4, 5 484
FQ209972.1 - 4, 5 484
FQ210003.1 - 4, 5 484
FQ210538.1 - 4, 5 484
FQ218232.1 - 4, 5 484
FQ218942.1 - 4, 5 484
FQ218960.1 - 4, 5 484
FQ218962.1 - 4, 5 484
FQ219069.1 - 4, 5 484
FQ219256.1 - 4, 5 484
FQ219272.1 - 4, 5 505
FQ219707.1 - 4, 5 484
M31363.1 - 4, 5 609

View Full Product Details