Gene Transcripts

Gene Symbol
Meig1
Entrez Gene ID
679834
Gene Name
meiosis/spermiogenesis associated 1
Gene Aliases
Chromosome Location
Chr. 17: 78813848 - 78829411 on Build Rnor_6.0
UniGene ID
Rn.46511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134882.1 NP_001128354.1 2 256
XM_006254276.2 XP_006254338.1 2 373
XM_006254278.2 XP_006254340.1 2 176
XM_008771888.1 XP_008770110.1 2 159

View Full Product Details