Gene Transcripts

Gene Symbol
Iffo2
Entrez Gene ID
641315
Gene Name
intermediate filament family orphan 2
Gene Aliases
Chromosome Location
Chr. 5: 158042587 - 158083311 on Build Rnor_6.0
UniGene ID
Rn.222020
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134703.1 NP_001128175.1 5 733
GenBank mRNA CV105369.1 - 2 219

View Full Product Details