Gene Transcripts

Gene Symbol
SMG5
Entrez Gene ID
23381
Gene Name
SMG5, nonsense mediated mRNA decay factor
Gene Aliases
EST1B, LPTS-RP1, LPTSRP1, SMG-5
Chromosome Location
Chr. 1: 156249224 - 156291498 on Build GRCh38
UniGene ID
Hs.516837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323614.1 NP_001310543.1 7 889
NM_001323615.1 NP_001310544.1 7 889
NM_001323616.1 NP_001310545.1 8 1021
NM_001323617.1 NP_001310546.1 8 856
NM_015327.2 NP_056142.2 7 828
XM_017000842.1 XP_016856331.1 3 774
GenBank mRNA AB029012.2 - 7 830
AB085691.1 - 7 756
AK092183.1 - 7 804
AY168922.1 - 7 682
AY336728.1 - 7 679
BC038296.1 - 7 829 4824487

View Full Product Details