Gene Transcripts
- Gene Symbol
- GAS5
- Entrez Gene ID
- 60674
- Gene Name
- growth arrest specific 5 (non-protein coding)
- Gene Aliases
- NCRNA00030, SNHG2
- Chromosome Location
- Chr. 1: 173863901 - 173867987 on Build GRCh38
- UniGene ID
- Hs.531856
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002578.2 |
|
9 |
371 |
|
GenBank mRNA |
BC038733.1 |
- |
7 |
1092 |
|
BC070486.1 |
- |
1 |
338 |
|
View Full Product Details