Gene Transcripts

Gene Symbol
HEBP2
Entrez Gene ID
23593
Gene Name
heme binding protein 2
Gene Aliases
C6ORF34B, C6orf34, PP23, SOUL
Chromosome Location
Chr. 6: 138404199 - 138415593 on Build GRCh38
UniGene ID
Hs.486589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001326380.1 NP_001313309.1 4 575
NM_001326381.1 NP_001313310.1 4 688
NM_014320.2 NP_055135.1 4 806
XM_017010631.1 XP_016866120.1 4 661
GenBank mRNA AF117616.1 - 4 784
AF411610.1 - 4 446
AK057928.1 - 4 721
AY427823.1 - 4 688
BC008205.1 - 4 670 3869026
BC010290.1 - 4 683 3445763
BC016806.1 - 4 786
BC016807.1 - 4 786
BC093037.1 - 4 669 30333215

View Full Product Details