Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001226.3 | NP_001217.2 | 7 | 804 | |
NM_032992.2 | NP_116787.1 | 4 | 537 | ||
NR_133012.1 | 6 | 727 | |||
GenBank mRNA | BC000305.2 | - | 7 | 752 | |
BC004460.1 | - | 7 | 752 | 2820468 | |
BM837197.1 | - | 4 | 483 | ||
BX380795.2 | - | 6 | 668 | ||
U20536.1 | - | 7 | 804 | ||
U20537.1 | - | 4 | 459 |