Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
Chromosome Location
Chr. 3: 122183683 - 122286503 on Build GRCh38
UniGene ID
Hs.435615
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000388.3 NP_000379.2 4 1615
NM_001178065.1 NP_001171536.1 4 1681
XM_005247837.2 XP_005247894.1 2 845
XM_006713789.3 XP_006713852.1 4 2196
XM_017007324.1 XP_016862813.1 5 3561
XM_017007325.1 XP_016862814.1 4 1587
GenBank mRNA BC104999.1 - 3 1267
BC112236.1 - 3 1267 8327704
D50855.1 - 3 1271
S83176.1 - 3 1243
U20759.1 - 4 1615
U20760.1 - 4 1681

View Full Product Details