Gene Transcripts

Gene Symbol
SAMHD1
Entrez Gene ID
25939
Gene Name
SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1
Gene Aliases
CHBL2, DCIP, HDDC1, MOP-5, SBBI88
Chromosome Location
Chr. 20: 36890882 - 36951843 on Build GRCh38
UniGene ID
Hs.580681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015474.3 NP_056289.2 10 1304
XM_005260384.3 XP_005260441.1 10 1277
XM_011528761.2 XP_011527063.1 10 1277
GenBank mRNA AB013847.1 - 10 1104
AB208944.1 - 10 1129
AF228421.1 - 10 1257
AK027811.1 - 10 1277
AK304187.1 - 10 1061
AK304795.1 - 11 1232
AK315169.1 - 10 1237
AL050267.1 - 10 1179
BC036450.2 - 10 1132 4816438

View Full Product Details