Gene Transcripts
- Gene Symbol
- HACL1
- Entrez Gene ID
- 26061
- Gene Name
- 2-hydroxyacyl-CoA lyase 1
- Gene Aliases
- 2-HPCL, HPCL, HPCL2, PHYH2
- Chromosome Location
- Chr. 3: 15560704 - 15601852 on Build GRCh38
- UniGene ID
- Hs.63290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284413.1 |
NP_001271342.1
|
12, 13 |
1566 |
|
NM_001284415.1 |
NP_001271344.1
|
11, 12 |
1467 |
|
NM_001284416.1 |
NP_001271345.1
|
10, 11 |
1401 |
|
NM_012260.3 |
NP_036392.2
|
13, 14 |
1647 |
|
NR_104315.1 |
|
11, 12 |
1468 |
|
GenBank mRNA |
AF161397.1 |
- |
13, 14 |
1267 |
|
AJ131753.1 |
- |
13, 14 |
1357 |
|
AK056977.1 |
- |
5, 6 |
641 |
|
AK301546.1 |
- |
12, 13 |
1283 |
|
AK301990.1 |
- |
10, 11 |
1136 |
|
AK302086.1 |
- |
11, 12 |
1184 |
|
BC001627.1 |
- |
13, 14 |
1365 |
3538159 |
BC007440.2 |
- |
11, 12 |
1173 |
|
View Full Product Details