Gene Transcripts

Gene Symbol
GIMAP2
Entrez Gene ID
26157
Gene Name
GTPase, IMAP family member 2
Gene Aliases
HIMAP2, IAN12, IMAP2
Chromosome Location
Chr. 7: 150685706 - 150693641 on Build GRCh38
UniGene ID
Hs.647071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015660.2 NP_056475.1 3 305
GenBank mRNA AL110151.1 - 3 284
BC013934.1 - 3 305 3950542
BC032345.1 - 3 281 5208411

View Full Product Details