Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016445.2 | NP_057529.1 | 2 | 335 | |
XM_011536644.2 | XP_011534946.1 | 2 | 309 | ||
GenBank mRNA | AF228603.1 | - | 2 | 246 | |
BC001226.1 | - | 2 | 279 | 3457876 | |
BC008056.2 | - | 2 | 189 | ||
CR976852.1 | - | 2 | 339 |