Gene Transcripts
- Gene Symbol
- PELP1
- Entrez Gene ID
- 27043
- Gene Name
- proline, glutamate and leucine rich protein 1
- Gene Aliases
- MNAR, P160
- Chromosome Location
- Chr. 17: 4671384 - 4704337 on Build GRCh38
- UniGene ID
- Hs.744899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278241.1 |
NP_001265170.1
|
10, 11 |
1374 |
|
NM_014389.2 |
NP_055204.3
|
9, 10 |
1292 |
|
GenBank mRNA |
AF547989.1 |
- |
9, 10 |
1066 |
|
AK226056.1 |
- |
9, 10 |
1072 |
|
AK291163.1 |
- |
9, 10 |
1092 |
|
AK293838.1 |
- |
10, 11 |
1161 |
|
AK299084.1 |
- |
9, 10 |
1280 |
|
AY882602.1 |
- |
9, 10 |
859 |
|
BC010457.2 |
- |
8, 9 |
820 |
|
BC069058.1 |
- |
9, 10 |
1066 |
6068755 |
U88154.1 |
- |
9, 10 |
973 |
|
View Full Product Details