Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318807.1 | NP_001305736.1 | 7 | 1773 | |
NM_014430.2 | NP_055245.2 | 6 | 1624 | ||
XM_011536659.2 | XP_011534961.1 | 6 | 1280 | ||
XM_017021221.1 | XP_016876710.1 | 6 | 3328 | ||
GenBank mRNA | AF190901.1 | - | 6 | 1668 | |
AF218586.1 | - | 4 | 802 | ||
AF544398.1 | - | 4 | 585 | ||
AK000387.1 | - | 4 | 579 | ||
BC035970.1 | - | 4 | 612 | 4716269 | |
CB162472.1 | - | 4 | 560 |