Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014508.2 | NP_055323.2 | 3, 4 | 555 | |
GenBank mRNA | AB208979.1 | - | 3, 4 | 520 | |
AF165520.1 | - | 3, 4 | 559 | ||
AK094570.1 | - | 3, 4 | 1869 | ||
AK313272.1 | - | 3, 4 | 559 | ||
BC011739.2 | - | 3, 4 | 555 | 4331363 | |
BC021080.1 | - | 3, 4 | 542 | ||
CR456394.1 | - | 3, 4 | 533 | ||
KC329528.1 | - | 3, 4 | 452 |