Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013258.4 | NP_037390.2 | 3 | 585 | |
NM_145182.2 | NP_660183.1 | 2 | 528 | ||
GenBank mRNA | AB023416.2 | - | 3 | 585 | |
AF184073.1 | - | 3 | 573 | ||
AF255794.1 | - | 2 | 516 | ||
AF310103.1 | - | 3 | 499 | ||
AF384665.1 | - | 3 | 552 | ||
AK000211.1 | - | 3 | 537 | ||
AK314422.1 | - | 3 | 577 | ||
BC004470.2 | - | 4 | 555 | 3678925 | |
BC013569.1 | - | 3 | 492 |