Gene Transcripts

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
Chromosome Location
Chr. 16: 67229389 - 67247522 on Build GRCh38
UniGene ID
Hs.95231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318202.1 NP_001305131.1 1, 2 324
NM_013241.2 NP_037373.2 1, 2 324
XM_011523043.2 XP_011521345.1 1, 2 540
XM_011523044.1 XP_011521346.1 1, 2 269
XM_011523045.2 XP_011521347.1 1, 2 34
GenBank mRNA AB041046.1 - 1, 2 294
AB209584.1 - 1, 2 266
AF113615.1 - 1, 2 230
AK304232.1 - 1, 2 280
AY192154.1 - 1, 2 212
BC033084.1 - 1, 2 269 5456031
CN302043.1 - 1, 2 326
KJ658417.1 - 1, 2 212

View Full Product Details