Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016433.3 | NP_057517.1 | 2, 3 | 274 | |
GenBank mRNA | AF209704.1 | - | 2, 3 | 170 | |
AK313457.1 | - | 2, 3 | 163 | ||
AY372530.1 | - | 2, 3 | 160 | ||
AY372531.1 | - | 2, 3 | 160 | ||
AY372532.1 | - | 2, 3 | 189 | ||
BC009932.2 | - | 2, 3 | 274 | 3959556 |