Gene Transcripts

Gene Symbol
TMEM69
Entrez Gene ID
51249
Gene Name
transmembrane protein 69
Gene Aliases
C1orf154
Chromosome Location
Chr. 1: 45688175 - 45694436 on Build GRCh38
UniGene ID
Hs.699548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016486.3 NP_057570.2 3 572
GenBank mRNA AF151063.1 - 3 551
AK024682.1 - 3 566
BC013608.1 - 3 554 3864518
BC034147.1 - 3 552 4655713
BC039718.2 - 2 475 5784175
BC046130.1 - 3 535 5787422
BC054041.1 - 1 286
BC064539.1 - 3 555 5725426
BC073836.1 - 2 472 5548608
BC121117.2 - 3 514 40120483
BC133692.1 - 3 513 40120480

View Full Product Details