Gene Transcripts

Gene Symbol
CEND1
Entrez Gene ID
51286
Gene Name
cell cycle exit and neuronal differentiation 1
Gene Aliases
BM88
Chromosome Location
Chr. 11: 787110 - 790126 on Build GRCh38
UniGene ID
Hs.22140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016564.3 NP_057648.2 2 711
GenBank mRNA AF235030.1 - 1 621
AK074547.1 - 2 708
BC034732.1 - 2 698 5179740

View Full Product Details