Gene Transcripts

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
Chromosome Location
Chr. 6: 110180421 - 110232220 on Build GRCh38
UniGene ID
Hs.428147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015891.2 NP_056975.1 3, 4 438
GenBank mRNA AB208903.1 - 3, 4 412
AF015044.1 - 3, 4 405
AF038392.1 - 3, 4 438
AF061241.1 - 3, 4 411
AK222721.1 - 3, 4 435
AK295055.1 - 3, 4 431
AK314601.1 - 3, 4 434
BC111061.1 - 4, 5 515
BC117145.1 - 3, 4 435 40125696
BC126114.1 - 3, 4 435 8991830

View Full Product Details