Gene Transcripts

Gene Symbol
SNX9
Entrez Gene ID
51429
Gene Name
sorting nexin 9
Gene Aliases
SDP1, SH3PX1, SH3PXD3A, WISP
Chromosome Location
Chr. 6: 157716046 - 157945077 on Build GRCh38
UniGene ID
Hs.191213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016224.4 NP_057308.1 16 1897
XM_005267015.2 XP_005267072.1 18 1975
XM_011535886.2 XP_011534188.1 13 1436
GenBank mRNA AB209736.1 - 10 3730
AF076957.1 - 11 1117
AF121859.1 - 16 1657
AF131214.1 - 16 1676
AF172847.1 - 16 1777
AK026637.1 - 13 1442
AK127345.1 - 13 1752
AK314360.1 - 16 1731
BC001084.2 - 7 698
BC005022.1 - 16 1804 3832234

View Full Product Details