Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001184728.2 | NP_001171657.1 | 4 | 549 | |
NM_001184729.2 | NP_001171658.1 | 5 | 717 | ||
NM_001184730.2 | NP_001171659.1 | 3 | 411 | ||
NM_015937.5 | NP_057021.2 | 5 | 717 | ||
NR_047691.1 | 5 | 677 | |||
NR_047692.1 | 4 | 620 | |||
NR_047693.1 | 4 | 616 | |||
NR_047694.1 | 4 | 539 | |||
NR_047695.1 | 2 | 310 | |||
XM_005260430.2 | XP_005260487.1 | 3 | 389 | ||
XM_005260432.2 | XP_005260489.1 | 4 | 511 | ||
GenBank mRNA | AB057724.1 | - | 5 | 627 | |
AF132940.1 | - | 5 | 621 | ||
AK075469.1 | - | 5 | 641 | ||
AK225517.1 | - | 5 | 647 | ||
AK293294.1 | - | 4 | 540 | ||
AK293686.1 | - | 4 | 462 | ||
AK296063.1 | - | 4 | 546 | ||
AK296139.1 | - | 4 | 472 | ||
AK299013.1 | - | 2 | 236 | ||
AK302093.1 | - | 3 | 369 | ||
AK311100.1 | - | 4 | 499 | ||
AK311416.1 | - | 3 | 361 | ||
AK311520.1 | - | 3 | 413 | ||
AK311522.1 | - | 4 | 528 | ||
AK316358.1 | - | 5 | 603 | ||
AL121742.1 | - | 5 | 620 | ||
AY358588.1 | - | 5 | 624 | ||
BC015022.1 | - | 5 | 646 | 3921680 | |
BC110892.1 | - | 5 | 625 | ||
BC136827.1 | - | 5 | 655 | 9020818 | |
BC136828.1 | - | 5 | 655 | 9020819 | |
BX537612.1 | - | 3 | 2055 |