Gene Transcripts

Gene Symbol
FBXO40
Entrez Gene ID
51725
Gene Name
F-box protein 40
Gene Aliases
FBX40
Chromosome Location
Chr. 3: 121593323 - 121630292 on Build GRCh38
UniGene ID
Hs.272564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016298.3 NP_057382.2 3 1162
GenBank mRNA AB033021.1 - 3 1366
AF204674.1 - 3 1068
AK314400.1 - 3 1084
BC109275.1 - 2 973 40028315
BC109276.1 - 2 973 40028318

View Full Product Details