Gene Transcripts

Gene Symbol
CEACAM5
Entrez Gene ID
1048
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
Gene Aliases
CD66e, CEA
Chromosome Location
Chr. 19: 41708585 - 41730433 on Build GRCh38
UniGene ID
Hs.709196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291484.2 NP_001278413.1 6 1618
NM_001308398.1 NP_001295327.1 6 1615
NM_004363.5 NP_004354.3 6 1618
XM_005258413.3 XP_005258470.2 6 1587
XM_011526322.1 XP_011524624.1 4 1053
XM_017026145.1 XP_016881634.1 6 1584
XM_017026146.1 XP_016881635.1 5 1332
GenBank mRNA AB852566.1 - 4 937
AK223101.1 - 6 1554
BC034671.1 - 6 1566 5184800
CR749337.1 - 4 660
M15042.1 - 6 1460
M16234.1 - 2 482
M17191.1 - 2 528
M29540.1 - 6 1585
X16455.1 - 6 1457

View Full Product Details