Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321073.1 | NP_001308002.1 | 4 | 532 | |
XM_011546094.1 | XP_011544396.1 | 4 | 424 | ||
XM_011546095.2 | XP_011544397.1 | 4 | 566 | ||
XM_011546097.2 | XP_011544399.1 | 5 | 593 | ||
XM_011546098.1 | XP_011544400.1 | 4 | 931 | ||
XM_011546099.1 | XP_011544401.1 | 4 | 424 | ||
XM_017028211.1 | XP_016883700.1 | 5 | 929 | ||
XM_017028212.1 | XP_016883701.1 | 6 | 985 | ||
XM_017028213.1 | XP_016883702.1 | 4 | 424 | ||
XM_017028214.1 | XP_016883703.1 | 5 | 926 | ||
XM_017028215.1 | XP_016883704.1 | 5 | 541 | ||
XM_017028216.1 | XP_016883705.1 | 4 | 931 | ||
XM_017028217.1 | XP_016883706.1 | 4 | 424 | ||
XM_017028218.1 | XP_016883707.1 | 5 | 593 | ||
GenBank mRNA | BC007257.1 | - | 4 | 502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000071.2 | NP_000062.1 | 4 | 525 | |
NM_001178008.2 | NP_001171479.1 | 4 | 532 | ||
NM_001178009.2 | NP_001171480.1 | 4 | 532 | ||
NM_001320298.1 | NP_001307227.1 | 5 | 526 | ||
XM_011529777.1 | XP_011528079.1 | 4 | 468 | ||
XM_011529781.1 | XP_011528083.1 | 5 | 533 | ||
XM_011529782.1 | XP_011528084.1 | 4 | 931 | ||
XM_017028489.1 | XP_016883978.1 | 5 | 928 | ||
XM_017028490.1 | XP_016883979.1 | 5 | 925 | ||
XM_017028491.1 | XP_016883980.1 | 4 | 468 | ||
XM_017028492.1 | XP_016883981.1 | 5 | 533 | ||
GenBank mRNA | AK294608.1 | - | 6 | 397 | |
AK301063.1 | - | 2 | 127 | ||
AK313691.1 | - | 4 | 430 | ||
AL137314.1 | - | 1 | 52 | ||
AL531222.3 | - | 4 | 516 | ||
BC000440.2 | - | 4 | 400 | 2820694 | |
BC010242.1 | - | 4 | 449 | 4154798 | |
BC011381.1 | - | 4 | 388 | 3869615 | |
BT007154.1 | - | 2 | 280 | ||
DC300040.1 | - | 4 | 525 | ||
DC315340.1 | - | 5 | 526 | ||
L14577.1 | - | 4 | 438 | ||
L19501.1 | - | 4 | 461 | ||
X82166.1 | - | 4 | 427 |