Gene Transcripts
- Gene Symbol
- CFL1
- Entrez Gene ID
- 1072
- Gene Name
- cofilin 1
- Gene Aliases
- CFL, HEL-S-15, cofilin
- Chromosome Location
- Chr. 11: 65854811 - 65858333 on Build GRCh38
- UniGene ID
- Hs.170622
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005507.2 |
NP_005498.1
|
3, 4 |
631 |
|
GenBank mRNA |
AK097690.1 |
- |
4, 5 |
1113 |
|
BC011005.2 |
- |
3, 4 |
459 |
4079579 |
BC012265.1 |
- |
3, 4 |
464 |
3862540 |
BC012318.1 |
- |
3, 4 |
463 |
3538185 |
BC018256.2 |
- |
3, 4 |
459 |
3448134 |
BC024975.1 |
- |
2, 3 |
166 |
|
BT006846.1 |
- |
2, 3 |
397 |
|
D00682.1 |
- |
2, 3 |
397 |
|
EU794603.1 |
- |
2, 3 |
397 |
|
U21909.1 |
- |
2, 3 |
397 |
|
X95404.1 |
- |
3, 4 |
448 |
|
View Full Product Details