Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243645.1 | NP_001230574.1 | 4 | 563 | |
NM_021914.7 | NP_068733.1 | 4 | 586 | ||
NM_138638.4 | NP_619579.1 | 4 | 728 | ||
NR_028130.1 | 4 | 585 | |||
NR_028131.1 | 4 | 474 | |||
XM_011536363.2 | XP_011534665.1 | 4 | 524 | ||
GenBank mRNA | AF087867.1 | - | 4 | 481 | |
AF134802.1 | - | 4 | 597 | ||
AF134803.1 | - | 4 | 491 | ||
AK314157.1 | - | 4 | 546 | ||
AL599958.1 | - | 4 | 562 | ||
BC011444.1 | - | 4 | 512 | 4245104 | |
BC022364.1 | - | 4 | 595 | 4730117 | |
BC022876.1 | - | 4 | 599 | 4109108 | |
BC025683.1 | - | 4 | 474 | ||
CB992605.1 | - | 4 | 428 | ||
DA298739.1 | - | 4 | 551 |