Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
54763
Gene Name
rhophilin associated tail protein 1
Gene Aliases
CT91, ODF6, RHPNAP1, ROPN1A, ropporin
Chromosome Location
Chr. 3: 123969006 - 123992170 on Build GRCh38
UniGene ID
Hs.567516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317774.1 NP_001304703.1 5 821
NM_001317775.1 NP_001304704.1 4 693
NM_017578.3 NP_060048.2 6 901
XM_011512933.2 XP_011511235.1 6 865
XM_011512934.2 XP_011511236.1 6 976
GenBank mRNA AF303889.2 - 6 901
BC132744.1 - 4 657 40146766
BC132746.1 - 4 658 40146768
BG219480.1 - 1 169
BG722939.1 - 4 653

View Full Product Details