Gene Transcripts

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
Chromosome Location
Chr. 1: 117860227 - 117930042 on Build GRCh38
UniGene ID
Hs.310809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135589.2 NP_001129061.1 9 1294
NM_017686.3 NP_060156.1 9 1294
XM_005270959.2 XP_005271016.1 9 1294
XM_011541621.2 XP_011539923.1 9 1294
XM_011541622.1 XP_011539924.1 9 1182
XM_011541623.2 XP_011539925.1 9 1294
XM_011541624.2 XP_011539926.1 10 1737
XM_017001531.1 XP_016857020.1 10 1316
XM_017001532.1 XP_016857021.1 10 1737
GenBank mRNA AK000149.1 - 9 1197
BC013132.1 - 9 1177 3448678

View Full Product Details