Gene Transcripts

Gene Symbol
PIGG
Entrez Gene ID
54872
Gene Name
phosphatidylinositol glycan anchor biosynthesis class G
Gene Aliases
GPI7, LAS21, MRT53, PRO4405, RLGS1930
Chromosome Location
Chr. 4: 499200 - 539921 on Build GRCh38
UniGene ID
Hs.7099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127178.2 NP_001120650.1 7 1361
NM_001289051.1 NP_001275980.1 7 1170
NM_001289052.1 NP_001275981.1 5 962
NM_001289053.1 NP_001275982.1 7 1919
NM_001289055.1 NP_001275984.1 8 1431
NM_017733.4 NP_060203.3 7 1361
NR_110293.1 7 1361
XM_005272284.2 XP_005272341.1 7 1595
XM_011513490.2 XP_011511792.1 7 1324
XM_011513491.1 XP_011511793.1 8 1391
XM_011513492.1 XP_011511794.1 2 245
XM_011513493.1 XP_011511795.1 7 1361
XM_011513494.2 XP_011511796.1 7 1361
XM_017008321.1 XP_016863810.1 7 1595
XM_017008322.1 XP_016863811.1 6 1453
XM_017008323.1 XP_016863812.1 8 1733
XM_017008324.1 XP_016863813.1 7 1595
XM_017008325.1 XP_016863814.1 8 1446
XM_017008326.1 XP_016863815.1 7 1361
GenBank mRNA AB162713.1 - 7 1331
AK000272.1 - 2 127
AK027465.1 - 1 80
AK074715.1 - 7 1133
AK074815.1 - 7 1340
AK296507.1 - 8 1422
AY358538.1 - 7 1303
BC001249.1 - 3 363
BC050262.1 - 7 1831
BC110878.1 - 5 904 5750347
BC144576.1 - 7 1318

View Full Product Details