Gene Transcripts

Gene Symbol
LGI2
Entrez Gene ID
55203
Gene Name
leucine rich repeat LGI family member 2
Gene Aliases
LGIL2
Chromosome Location
Chr. 4: 24991960 - 25030792 on Build GRCh38
UniGene ID
Hs.12488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018176.3 NP_060646.2 8 1234
GenBank mRNA AB067503.1 - 8 1127
AF467955.1 - 8 1135
AJ487516.1 - 8 1135
AJ487958.1 - 8 1157
AK001537.1 - 1 177
AL833647.1 - 7 925
BC101759.1 - 8 1234 8069265
BC101761.1 - 8 1234 8069267

View Full Product Details