Gene Transcripts

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
Chromosome Location
Chr. 4: 168216291 - 168318807 on Build GRCh38
UniGene ID
Hs.591710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017631.5 NP_060101.3 28 4014
XM_011532103.2 XP_011530405.1 28 4014
GenBank mRNA BC038115.2 - 28 3959
BX648655.1 - 8 856

View Full Product Details