Gene Transcripts

Gene Symbol
RAB20
Entrez Gene ID
55647
Gene Name
RAB20, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 13: 110523066 - 110561737 on Build GRCh38
UniGene ID
Hs.743563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017817.2 NP_060287.1 2 705
GenBank mRNA AJ272065.1 - 2 487
AK000436.1 - 2 692
AK000446.1 - 2 692
BC026025.1 - 2 701 4636491

View Full Product Details