Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018085.4 | NP_060555.2 | 22 | 2974 | |
GenBank mRNA | AB033018.1 | - | 5 | 588 | |
AF161391.1 | - | 5 | 628 | ||
AF410465.1 | - | 22 | 2924 | ||
AK001264.1 | - | 8 | 1146 | ||
AK027532.1 | - | 13 | 1838 | ||
AK074740.1 | - | 13 | 1855 | ||
AL834323.1 | - | 7 | 1031 | ||
BC003604.1 | - | 11 | 1710 |