Gene Transcripts

Gene Symbol
NECAP2
Entrez Gene ID
55707
Gene Name
NECAP endocytosis associated 2
Gene Aliases
Chromosome Location
Chr. 1: 16440672 - 16460090 on Build GRCh38
UniGene ID
Hs.437385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145277.1 NP_001138749.1 2 236
NM_001145278.1 NP_001138750.1 2 158
NM_018090.4 NP_060560.1 2 236
XM_011541767.2 XP_011540069.1 3 308
XM_017001721.1 XP_016857210.1 2 184
GenBank mRNA AK001282.1 - 2 168
AK023545.1 - 2 172
AK024468.1 - 2 150
AK223344.1 - 2 184
AK300764.1 - 2 182
AK302227.1 - 2 104
BC017014.1 - 2 167 3848563
BC018914.2 - 2 170 4299890
BM751287.1 - 2 236

View Full Product Details