Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018448.4 | NP_060918.2 | 10 | 2209 | |
GenBank mRNA | AB020636.2 | - | 10 | 2111 | |
AF157326.1 | - | 10 | 2126 | ||
AK027404.1 | - | 4 | 844 | ||
AK074922.1 | - | 7 | 1383 | ||
AK292456.1 | - | 12 | 2048 | ||
AK314358.1 | - | 10 | 2097 | ||
AL136810.1 | - | 10 | 2121 | ||
BC026220.2 | - | 8 | 1560 | ||
BC050341.1 | - | 10 | 1905 | 5265409 |