Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
Chromosome Location
Chr. 12: 16548372 - 16610233 on Build GRCh38
UniGene ID
Hs.504908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001395.2 NP_001001395.1 2 348
NM_001243609.1 NP_001230538.1 3 415
NM_001243610.1 NP_001230539.1 3 187
NM_001243611.1 NP_001230540.1 2 584
NM_001243612.1 NP_001230541.1 2 272
NM_001243613.1 NP_001230542.1 3 286
NM_018640.4 NP_061110.2 2 606
NR_045012.1 3 415
NR_045013.1 3 415
NR_045014.1 2 348
XM_006719110.3 XP_006719173.1 3 423
XM_006719111.2 XP_006719174.1 2 466
XM_011520762.1 XP_011519064.1 3 314
XM_011520763.1 XP_011519065.1 3 221
XM_017019643.1 XP_016875132.1 2 569
GenBank mRNA AB044745.1 - 2 352
AB044746.1 - 4 253
AF258348.1 - 3 204
AK095595.1 - 2 224
AK294474.1 - 2 272
AK294651.1 - 2 261
AK294909.1 - 3 286
AK308594.1 - 3 326
AK308826.1 - 4 291
AK308896.1 - 3 227
AK312418.1 - 2 490
AK316021.1 - 2 208
BC017777.1 - 2 161
BC026311.1 - 2 502 4799836
BC050085.1 - 3 188 5274811
BI553855.1 - 2 244
DA780743.1 - 2 349

View Full Product Details