Gene Transcripts

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
Chromosome Location
Chr. 5: 141398128 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018925.2 NP_061748.1 1 1198
NM_032099.1 NP_115270.1 1 1198
GenBank mRNA AF152334.1 - 1 1198
AF152521.1 - 1 1198
BC093877.1 - 1 1289 7939722
BC093879.1 - 1 1289 7939724

View Full Product Details