Gene Transcripts

Gene Symbol
XAB2
Entrez Gene ID
56949
Gene Name
XPA binding protein 2
Gene Aliases
HCNP, HCRN, NTC90, SYF1
Chromosome Location
Chr. 19: 7619525 - 7629553 on Build GRCh38
UniGene ID
Hs.9822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020196.2 NP_064581.2 3 242
GenBank mRNA AB026111.1 - 3 231
AF226051.1 - 3 242
AF258567.1 - 3 237
AF272147.1 - 3 252
AK074035.1 - 1 435
BC007208.1 - 3 224 2823235

View Full Product Details