Gene Transcripts

Gene Symbol
NLGN4X
Entrez Gene ID
57502
Gene Name
neuroligin 4, X-linked
Gene Aliases
ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
Chromosome Location
Chr. X: 5890026 - 6228882 on Build GRCh38
UniGene ID
Hs.21107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282145.1 NP_001269074.1 3 1135
NM_001282146.1 NP_001269075.1 2 810
NM_020742.3 NP_065793.1 2 645
NM_181332.2 NP_851849.1 2 828
XM_005274564.2 XP_005274621.1 2 822
XM_005274565.2 XP_005274622.1 2 805
XM_005274566.4 XP_005274623.1 2 749
XM_006724504.3 XP_006724567.1 2 690
XM_011545547.2 XP_011543849.1 2 824
XM_011545548.2 XP_011543850.1 2 988
XM_017029690.1 XP_016885179.1 2 634
XM_017029691.1 XP_016885180.1 2 824
XM_017029692.1 XP_016885181.1 2 749
XM_017029693.1 XP_016885182.1 2 634
GenBank mRNA AB033086.1 - 2 645
AF376803.1 - 2 645
AK055471.1 - 3 868
AK291036.1 - 2 682
AX773938.1 - 2 645
AY358562.1 - 1 173
BC034018.1 - 2 611 4702963

View Full Product Details