Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080679.2 | NP_542410.2 | 26 | 2161 | |
NM_080680.2 | NP_542411.2 | 29 | 2482 | ||
NM_080681.2 | NP_542412.2 | 27 | 2224 | ||
XM_011514299.2 | XP_011512601.1 | 24 | 1773 | ||
XM_011514300.2 | XP_011512602.1 | 23 | 1593 | ||
XM_011514302.2 | XP_011512604.1 | 22 | 1794 | ||
XM_017010250.1 | XP_016865739.1 | 30 | 2747 | ||
XM_017010251.1 | XP_016865740.1 | 22 | 1611 | ||
GenBank mRNA | J04974.1 | - | 2 | 67 | |
L18987.1 | - | 26 | 1843 |