Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
57687
Gene Name
vesicle amine transport 1-like
Gene Aliases
Chromosome Location
Chr. 16: 77788564 - 77980107 on Build GRCh38
UniGene ID
Hs.461405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020927.2 NP_065978.1 4 805
GenBank mRNA AB046796.1 - 4 775
AK290153.1 - 4 783
AL834240.1 - 3 420
BC033537.1 - 4 783 4821265

View Full Product Details