Gene Transcripts

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
Chromosome Location
Chr. 14: 73058436 - 73121675 on Build GRCh38
UniGene ID
Hs.531106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021239.2 NP_067062.1 8 980
XM_011537044.2 XP_011535346.1 9 1082
XM_011537045.2 XP_011535347.1 8 892
GenBank mRNA AK026107.1 - 8 960
AK094697.1 - 7 824
AL832314.1 - 8 951
BC062440.1 - 8 984
BC083496.1 - 8 946
BC111066.1 - 8 944
BC113389.1 - 7 760
BC113391.1 - 7 760
BC136775.1 - 8 817 9020765
BC136776.1 - 8 817 9020766
BC144407.1 - 9 885 9052928
BX647116.1 - 8 933

View Full Product Details