Gene Transcripts

Gene Symbol
NIF3L1
Entrez Gene ID
60491
Gene Name
NGG1 interacting factor 3 like 1
Gene Aliases
ALS2CR1, CALS-7, MDS015
Chromosome Location
Chr. 2: 200889327 - 200903932 on Build GRCh38
UniGene ID
Hs.145284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136039.2 NP_001129511.1 4 715
NM_001142355.1 NP_001135827.1 4 664
NM_001142356.1 NP_001135828.1 3 650
NM_021824.3 NP_068596.2 4 867
XM_005246742.3 XP_005246799.1 4 930
XM_011511582.1 XP_011509884.1 4 851
XM_011511583.1 XP_011509885.1 4 1595
XM_011511584.1 XP_011509886.1 4 1413
XM_011511585.1 XP_011509887.1 4 1362
XM_017004630.1 XP_016860119.1 4 1112
GenBank mRNA AB038949.1 - 4 828
AF060513.1 - 4 624
AF182416.1 - 4 789
AF283538.1 - 4 670
AK023378.1 - 4 813
AK225905.1 - 4 2190
AK314709.1 - 4 669
AL834430.1 - 3 490
AY251943.1 - 3 653
AY251944.1 - 3 653
BC007654.2 - 3 651 3346099

View Full Product Details