Gene Transcripts

Gene Symbol
RBKS
Entrez Gene ID
64080
Gene Name
ribokinase
Gene Aliases
RBSK, RK
Chromosome Location
Chr. 2: 27781364 - 27890396 on Build GRCh38
UniGene ID
Hs.11916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287580.1 NP_001274509.1 4, 5 1464
NM_022128.2 NP_071411.1 3, 4 345
GenBank mRNA AJ404857.1 - 3, 4 305
AK300989.1 - 3, 4 299
BC017425.1 - 3, 4 347 4694872
BX648909.1 - 4, 5 1435

View Full Product Details